Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Glassware - Round Bottom Flasks
SDP

For use with StarFish™ multi-experiment heating and stirring work station

Supplier:  Heidolph™ 015890740

 View more versions of this product

Catalog No. 05-404-101


Add to Cart

Description

Description

  • Choice of sizes: 5 mL, 10 mL, 25 mL, 50 mL, 100 mL, and 250 mL
Specifications

Specifications

1.69 oz.
50 mL
StarFish Multi-Experiment Heating and Stirring Work Station
14/23
14/23
With 14/23 socket and 14/23 sidearm
Glass
2
14/23
Round
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit