Learn More
Heidolph™ Heat-On™ Multi-Well Holder Insert
GREENER_CHOICESDP
Insert for Heidolph Heat-On Multi-Well Holders when used with Hei-PLATE magnetic stirring hotplates.
Supplier:  Heidolph™ 015882130
 View more versions of this product
Catalog No. 05-404-169
Add to Cart
Description
Description
  • Inserts are available for 10 mL, 25 mL, 50 mL, 100 mL and 150 mL flasks
  • Flask inserts also feature cut-away sides for use with two or three neck flasks
  • Multi-tube inserts are available for 16 mm, 20 mm, and 24 mm tubes
  • Available with fluoropolymer coating or anodized finish
Specifications
Specifications
4 x 24mm fluoropolymer coated tube of Heat-On™ system
4 x 24 mm Tubes
Fluoropolymer
Multi-Well Holder, Hei Series Hotplate
Heat-On Insert
Product Suggestions
Product Suggestions