Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-VAP Industrial B Rotary Evaporator
SDP

Features identical specifications as the Hei-VAP Industrial, but for limited budgets and comes without certain safety aspects.

Supplier:  Heidolph™ 036072055

 View more versions of this product

Catalog No. 13-890-112


Add to Cart

Description

Description

  • Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180 °C
  • Receiver cassettes and additional PMMA door housing protect against threat of glassware breakage
  • Comes as standard with integrated refill water system, spillover prevention and a release valve at the bottom
  • Optional Base Cart is fully-maneuverable with removable panels for storage of vacuum pump, glassware, solvents or accessories
  • Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ)
  • Large touch screen control panel with illuminated displays for all process parameters, programmable ramps
  • Features identical specifications as the Hei-VAP Industrial, but for limited budgets and comes without these safety aspects:

    • Metal frame and safety glass guard hood covering the bath
    • High-impact PMMA door covering the glassware assembly
    • High-impact PMMA latching doors which cover the receiving flask cassettes
    Specifications

    Specifications

    230 V
    20°C to 180°C
    RC Glassware: one ascending condenser, one 20 L evaporating flask, two 10 L receiving flask
    Motor Lift Rotary Evaporator
    6 to 160 rpm
    Recommended for All Standard Distillations
    NRTL
    Glassware RC
    Product Suggestions

    Product Suggestions

    Safety and Handling

    Safety and Handling

    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    SDS
    Documents

    Documents

    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit