Learn More
Heidolph™ Hei-VAP Industrial Rotary Evaporator
Hei-VAP Industrial Rotary Evaporator is perfectly designed for a great deal of different distilling processes, from standard evaporation without vacuum control up to complex distillation processes with vacuum control.
Manufacturer:  Heidolph™ 036072045
 View more versions of this product
Catalog No. 13-890-109
Add to cart
Description
Description
  • Excellent user safety with high-impact transparent PMMA door Non-fogging safety glass and metal frame guard hood provides excellent user protection
  • The evaporation flask is illuminated during operation for increased visibility
  • Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180 °C
  • Receiver cassettes and additional PMMA door housing protect against threat of glassware breakage
  • Comes as standard with integrated refill water system, spillover prevention and a release valve at the bottom
  • Optional Base Cart is fully-maneuverable with removable panels for storage of vacuum pump, glassware, solvents or accessories
  • Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ)
  • Large touch screen control panel with illuminated displays for all process parameters, programmable ramps
  • Specifications
    Specifications
    Hei-VAP Industrial S Rotary Evaporator
    NRTL
    KC Glassware: one 20 L evaporating flask and two 10 L receiving flasks
    20°C to 180°C
    Glassware KC
    Condensate Trap for Low-Boiling Solvents; Cooling by Dry-Ice
    motor lift
    6 to 160 rpm
    230 V