Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-VAP Industrial Rotary Evaporator
SDP

Hei-VAP Industrial Rotary Evaporator is perfectly designed for a great deal of different distilling processes, from standard evaporation without vacuum control up to complex distillation processes with vacuum control.

Supplier:  Heidolph™ 036072035

 View more versions of this product

Catalog No. 13-890-107


Add to Cart

Description

Description

  • Excellent user safety with high-impact transparent PMMA door Non-fogging safety glass and metal frame guard hood provides excellent user protection
  • The evaporation flask is illuminated during operation for increased visibility
  • Universal heating bath accommodates water or other bath fluids allowing for temperature settings up to 180 °C
  • Receiver cassettes and additional PMMA door housing protect against threat of glassware breakage
  • Comes as standard with integrated refill water system, spillover prevention and a release valve at the bottom
  • Optional Base Cart is fully-maneuverable with removable panels for storage of vacuum pump, glassware, solvents or accessories
  • Certification according to GMP available for this model: validation for installation (IQ) and operating qualification (OQ)
  • Large touch screen control panel with illuminated displays for all process parameters, programmable ramps
  • Specifications

    Specifications

    230 V
    20°C to 180°C
    A2C Glassware: one ascending, one descending condenser and one expansion vessel, one 20 L evaporating flask, two 10 L receiving flasks
    Motor Lift Rotary Evaporator
    6 to 160 rpm
    For Low-Boiling Solvents and Highest Recovery Rates
    NRTL
    Glassware A2C
    Product Suggestions

    Product Suggestions

    Safety and Handling

    Safety and Handling

    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
    SDS
    Documents

    Documents

    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit