Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Powder Flasks Available on GSA/VA Contract for Federal Government customers only.
Powder flasks for use with Heidolph Hei-VAP series rotary evaporators
Manufacturer: Heidolph North America 036302220
 View more versions of this product
Catalog No. 13-878-679
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 500mL
    For Use With (Equipment) Hei-VAP rotary evaporator
    Product Type Powder Drying Flask
    Taper 29/32
  • Description & Specifications
Related Products
Product Content Feedback