Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Spiders
SDP

For use with Heidolph Hei-VAP series rotary evaporators

Supplier:  Heidolph™ 036302690

 View more versions of this product

Catalog No. 13-878-726


Add to Cart

Description

Description

  • Processes multiple samples at the same time
  • Choice of models: NS 14.5 spider with 6, 12 or 20 distillation sleeves (20 mL) or NS 24 spider with 5 flasks (50 or 100 mL)
  • Available in NS 24/40 or 29/32 joint size

Specifications

Specifications

Spider Flasks
6 Tubes
29/32
Hei-VAP Rotary Evaporator
6/Pk.
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit