Learn More
Heidolph™ Hei-Vap™ Rotary Evaporators - Advantage - Complete Packages Available on GSA/VA Contract for Federal Government customers only.
For standard distillations with regulated vacuum control (and chiller when applicable)
Manufacturer:  Heidolph North America 036020440
 View more versions of this product
Catalog No. 13-876-176
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
145.4 oz., 4300mL
115V
145.4 oz.
20 to 270rpm
Standard distillation with regulated vacuum control
Hei-VAP advantage G3B/ML, rotavac valve tec vacuum pump, manual vacuum controller, tube set
Large dial control
50/60Hz
24/40
4300mL
20° to 100°C
Advantage 5 Rotary Evaporator Package
68° to 212°F
115V 50/60Hz