Learn More
Heidolph™ Hei-Vap™ Rotary Evaporators - Precision - Complete Packages Available on GSA/VA Contract for Federal Government customers only.
For most demanding applications
Manufacturer:  Heidolph North America 036021120
 View more versions of this product
Catalog No. 13-900-116
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
145.4 oz., 4300mL
115V
145.4 oz.
20 to 270rpm
Most demanding applications
Hei-VAP precision G1B/HL, Hei-VAP rotavac vario tec pump, Hei-VAP vapor temperature sensor, tube set
115V 50/60Hz
50/60Hz
24/40
4300mL
20° to 100°C
Precision 13 Hei-Vap Rotary Evaporator Package
68° to 212°F