Learn More
Heidolph™ Hei-Vap™ Rotary Evaporators - Value “The Collegiate” - Complete Packages
Standard equipment for the lab
Manufacturer:  Heidolph North America 036020050
 View more versions of this product
Catalog No. 13-876-157
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
145.4 oz., 4300mL
115V
145.4 oz.
20 to 270rpm
Standard distillation, standard equipment for the lab
Hei-VAP value G3B, B-169 water aspirator, manual vacuum control, tube set
115V 50/60Hz
50/60Hz
24/40
4300mL
20° to 100°C
Value 5 Hei-Vap Rotary Evaporator Package
68° to 212°F