Learn More
Heidolph™ Immersion Blenders - SilentCrusher M Accessories - Generators
Generators used with Heidolph SilentCrusher M blender
Manufacturer:  Heidolph North America 036300730
 View more versions of this product
Catalog No. 13-889-901
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
3 to 50mL
Generator
SilentCrusher M blender
Stainless Steel
24dBA
26000rpm
8cm
Ideal for applications under vacuum down to 20mbar or pressure up to 6bar
3.15 in.
8 F
<0.04cm