Learn More
Heidolph™ Immersion Blenders - SilentCrusher M Accessories - Generators
Generators used with Heidolph SilentCrusher M blender
Manufacturer:  Heidolph North America 036300830
 View more versions of this product
Catalog No. 13-900-805
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
100 to 2000mL
Generator
SilentCrusher M blender
Stainless Steel
73dBA
20000rpm
18cm
Ideal for applications under vacuum down to 20mbar or pressure up to 6bar
7.08 in.
22 F
<0.04cm