Learn More
Heidolph™ Immersion Blenders - SilentCrusher M Accessories - Generators Available on GSA/VA Contract for Federal Government customers only.
Generators used with Heidolph SilentCrusher M blender
Manufacturer:  Heidolph North America 036300780
 View more versions of this product
Catalog No. 13-900-800
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
25 to 250mL
Generator
SilentCrusher M blender
Stainless Steel
62dBA
26000rpm
11.5cm
Ideal for applications under vacuum down to 20mbar or pressure up to 6bar
4.52 in.
12 DG
<0.2cm