Learn More
Heidolph™ Immersion Blenders - SilentCrusher M Accessories - Generators Available on GSA/VA Contract for Federal Government customers only.
Generators used with Heidolph SilentCrusher M blender
Manufacturer:  Heidolph North America 036306020
 View more versions of this product
Catalog No. 13-889-464
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
0.8 to 30mL
Generator
SilentCrusher M blender
Stainless Steel
73dBA
26000rpm
6cm
Ideal for applications under vacuum down to 20mbar or pressure up to 6bar
2.36 in.
6 F
<0.04cm