Learn More
Heidolph™ Hei-VAP Industrial and Laborota 20 Rotary Evaporator Accessories
Accessories for use with Heidolph Hei-VAP Industrial and Laborota 20 rotary evaporators only
Manufacturer:  Heidolph™ 036303060
 View more versions of this product
Catalog No. 13-878-858
Add to cart
Description
Description
  • The vacuum sensor controls and displays the actual vacuum controls in addition the maximum permissible pressure.
  • The vacuum valve controls vacuum.
  • Vapor temperature sensor measure the temperature of vapors in the condenser.
  • Tubing set is 6 meter of vacuum and 10 meter of enforced cooling tube including hose clamps.
Specifications
Specifications
Vacuum Pressure Sensor
Hei-VAP Industrial and Laborota 20 Rotary Evaporator
Vacuum Pressure Sensor
Standard for LR 20 compact, safety and automatic