Learn More
Heidolph™ Rotary Evaporator Accessories - Interchangeable Glassware
Five condenser assemblies allow customization of evaporator glassware for any application
Supplier: Heidolph™ 036301930
Description
- All glassware can be interchanged among rotary evaporator models
- Each set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
- All sets are available with standard or plastic safety-coated glassware (halar coating for G5B and abcite coating for G1B/G3B/G6B)
- Evaporating flasks and vapor tubes with the standard ground joint NS 24/40 are also available with NS 29/32 ground joints
Specifications
Screwing for G6 Condenser | |
Rotary Evaporator |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov