Heidolph™ Lara Controlled Lab Reactor Accessory, Probes
Temperature probes
Manufacturer: Heidolph North America 015907068
 View more versions of this product
Catalog No. 05-406-462
  Check Availability
Add to Cart
    Specifications
    Product Type Temperature Probe
    For Use With (Equipment) Lara controlled lab reactor
    Length (Metric) 400mm
    Material PTFE
  • Description & Specifications
Related Products
Product Content Feedback