Learn More
Heidolph™ Mechanical Overhead Stirrers
Accomplish the most demanding tasks while providing the highest safety and increased performance life
Manufacturer:  Heidolph North America 036090070
 View more versions of this product
Catalog No. 13-880-113
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
60Hz
8.3 in.
40 to 2000rpm
70/37w
3.2 in.
8.2cm
17.6cm
2
32° to 104°F
1352.5 oz.
115V 60Hz
0° to 40°C
21.1cm
115V
7.2 lb.
3.3kg
6.9 in.
Designed to mix and disperse media that require non-reproducible results of high-viscosity applications under high speeds, these models are ideal for standard stirring tasks
10mm Chuck
520
40000mL
100000
Accessories