Learn More
Heidolph™ MR Series Magnetic Stirrer Hotplates Available on GSA/VA Contract for Federal Government customers only.
For smooth to intense mixing and heating of low-viscosity fluids and an excellent choice for decomposing organic and inorganic substances.
Manufacturer:  Heidolph North America 036110580
 View more versions of this product
Catalog No. 13-889-336
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Stir: 20L; Load: 25kg
9.4cm
17.3cm
600 - 820w
2.6kg
27.7cm
115V
5.68 in.
115V 60Hz
14.5cm
3.68 in.
6.81 in.
MR Hei-End Standard
5.7 lb.
10.87 in.
30 to 1400rpm
60Hz
300°C
6.81 x 10.87 x 3.68 in. (17.3 x 27.7 x 9.4cm)