Learn More
Heidolph™ Hei-PLATE Mix 'n' Heat Magnetic Stirrer Hotplate Accessory, Temperature Sensor
Temperature Sensor for measuring and regulating the temperature of the medium, providing precise temperature control.
Supplier: Heidolph™ 036110350
Description
Single Sensors
- Choice of stainless steel or glass-coated PT1000
- 4-pin connection
- For use with Heidolph Hei-PLATE Expert and Hei-PLATE Ultimate magnetic stirring hotplates
Specifications
Silver/White | |
For measuring and regulating the temperature of the medium, providing precise temperature control | |
PT1000, Stainless Steel | |
Stainless Steel |
Hei-PLATE Mix 'n' Heat Core+, Hei-PLATE Mix 'n' Heat Expert, Hei-PLATE Mix 'n' Heat Ultimate | |
Stainless Steel Probe | |
Temperature Sensor |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov