Learn More
Heidolph™ PC Interface, Analog for RZR Overhead Stirrer
Accessory for use with RZR Overhead Stirrers
Manufacturer:  Heidolph™ 036300630
Catalog No. 13-880-157
Add to cart
Description
Description
  • For use with RZR overhead stirrers
  • Integrates stirrer in process automation
Specifications
Specifications
RZR Overhead Stirrer
Analog Interface