Learn More
Heidolph™ Peristaltic Pump Drives Available on GSA/VA Contract for Federal Government customers only.
Smooth start operation prevents spills and media splashing
Manufacturer:  Heidolph North America 036150170
 View more versions of this product
Catalog No. 13-889-878
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
±0.5%
25.6cm
8.85 in.
50/60Hz
PD5201
0° to 40°C
150w
115V
Digital
10.07 in.
115V 50/60Hz
22.5cm
Adaptor for multichannel operation
5 to 120rpm
10000
6.53 in.
Electronically commutated direct current (DC) motor
0.3 to 790/0.005 to 320mL/min.