Learn More
Heidolph™ Polymax Wave Motion Platform Shakers Available on GSA/VA Contract for Federal Government customers only.
Platform lab shaker with wave motion accepts load capacities of 5 or 10kg
Manufacturer:  Heidolph North America 036130320
 View more versions of this product
Catalog No. 13-878-461
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polymax 1040
0 to 50°C
11.03 lb.
10°
7.6 in.
12.5 in.
37.5cm
17.63 lb.
115w
Acoustic
115V 60Hz
2 to 50rpm
115V
5kg
1 to 120 min.
14.3 in.
19.5cm
32cm
8kg
60Hz
14.3 x 12.5 x 7.6 in. (37.5 x 32 x 19.5cm)