Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Pump Tubing - PharMed™ Extension Tube Available on GSA/VA Contract for Federal Government customers only.

For multi-channel pumps (PD5001/5101/5201)

Manufacturer:  Heidolph North America 036303910

 View more versions of this product

Catalog No. 13-889-840


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Thermoplastic Elastomer
Recommended
0.1 in.
0.035 in.
Recommended
Yes
-76 to 275°F
Yes
22.9 ft. Water
Recommended
2.69mm
Extension Pump Tubing
FDA 21 CFR 177.2600, USP 23 class VI, GLP, pharmacopoea and europaea
Not recommended
0.9mm
Very low
Opaque
-60 to 135°C
Multi channel pump (PD5001, PD5101, PD5201)
7m Water
0.89mm
0.035 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit