Learn More
Heidolph™ Radleys Carousel Zinc Plated Wire Rack
Carousel zinc plated wire rack for use with Carousel 12 Plus Reaction Station™
Supplier: Heidolph™ 015860450
Description
Choice of size for 36 x 24 mm, 40 x 24 mm, and 80 x 24 mm tubes.
Specifications
Silver | |
Zinc Plated | |
Non-sterile | |
Carousel 12 Plus Reaction Station™ |
Tubes (80 x 24 mm) | |
80 | |
Wire Rack |