Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Radleys Carousel Zinc Plated Wire Rack
SDP

Carousel zinc plated wire rack for use with Carousel 12 Plus Reaction Station™

$214.00

Specifications

Product Type Wire Rack
Color Silver
Material Zinc Plated
Sterility Non-sterile
For Use With (Equipment) Carousel 12 Plus Reaction Station™
View More Specs

Products 2
Catalog Number Mfr. No. Holds Price Quantity & Availability  
Catalog Number Mfr. No. Holds Price Quantity & Availability  
05-403-772
SDP
View Documents
Heidolph™
015860430
Tubes (36 x 24 mm)
Each for $214.00
Add to Cart
 
05-403-773
SDP
View Documents
Heidolph™
015860440
Tubes (40 x 24 mm) Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. N/A
Description

Description

Choice of size for 36 x 24 mm, 40 x 24 mm, and 80 x 24 mm tubes.

Specifications

Specifications

Wire Rack
Zinc Plated
Carousel 12 Plus Reaction Station™
Silver
Non-sterile
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit