Learn More
Heidolph™ Radleys GreenHouse Blowdown Evaporator™ Accessories: Blowdown Pin Plates
SDP
For use with GreenHouse Blowdown Evaporator™
Manufacturer:  Heidolph™ 015860660
 View more versions of this product
Catalog No. 05-403-795
Add to cart
Description
Description
Interchangeable pin plates with either 8 or 24 hollow blowdown pins deliver an equal flow of gas to each tube, vial or well
Specifications
Specifications
Heidolph Radleys GreenHouse Blowdown Evaporator™
Blowdown Pin Plate, 6 x 4