Learn More
Heidolph™ Radleys GreenHouse Plus Parallel Synthesizer™ Accessories - Septum Mats
SDP
For use with GreenHouse Plus Parallel Synthesizer™
Supplier:  Heidolph™ 015880890
 View more versions of this product
Catalog No. 05-403-981
Add to Cart
Description
Description
  • Allows additions and withdrawals through sealing mats
  • Silicone Septum Mat: The silicone exhibits excellent resistant to heat and cold as well as offering good resistance to a broad range of solvents and solvent vapours. Can be repeatedly pierced whilst maintaining a gas tight seal.
  • Pre-Slit PTFE Faced Silicone Septum Mat: Manufactured from chromatographically clean silicone elastomer, with a thin PTFE film on both sides and 24 pre-slit positions enabling use of blunt syringe needles.
  • Specifications
    Specifications
    GreenHouse Plus Parallel Synthesizer™
    Septum Mat, PTFE-Faced, Pre-Slit