Learn More
Heidolph™ Radleys Jacketed Vessel Kit
GSA_VASDP
Radleys jacketed vessel kit for use with Reactor-Ready™ and Reactor-Ready™ Duo lab reactors
Manufacturer:  Heidolph™ 015950112
 View more versions of this product
Catalog No. 05-405-264
Add to cart
Description
Description
  • These cost effective vessel kits include the vessel and accessories you need to get started.
  • The Pt100 probe and anchor stirrer included in each kit are matched to the vessel size.
  • Each vessel kit includes reaction vessel, PTFE anchor stirrer, side arm couplings, Pt100 PTFE temperature probe with LEMO connector, and PTFE temperature probe adapter.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
446°F
5000 mL
Reactor-Ready™ and Reactor-Ready™ Duo Lab Reactors
Vacuum jacketed
Jacketed Vessel Kit