Learn More
Heidolph™ Radleys SpeediFlow™ Adapter Heads
SpeediFlow™ adapter heads for use with Carousel Work-up Station™
Manufacturer:  Heidolph™ 015860060
 View more versions of this product
Catalog No. 05-403-735
Add to cart
Description
Description
  • The SpeediFlow is a hand held pressurizer which fits into the top of your plastic column, to push the mobile phase through the column.
  • Features a mechanically expandable Viton O-ring, operated by a trigger with an integral locking device, for unsupported gas tight operation
  • SpeediFlow features a safety relief valve that vents at 6 to 8 psi, preventing accidental over-pressure
  • SpeediFlow Booster increases flow rates to speed up your work-ups.
  • The SpeediFlow includes a range of optional adapters to fit all standard plastic column sizes including 15 ml, 25 ml, 70 ml, and 150 ml, plus the Whatman AutoCup™. Thus allowing the SpeediFlow to be used in other SPE or filtration applications.
  • Specifications
    Specifications
    25 mL Columns
    Adapter Head