Learn More
Heidolph™ Starfish Workstation Accessories - MonoBlock and PolyBlock Inserts
SDP
For use with Heidolph MonoBlock (5 x 250 mL) and Polyblock (1 x 250 mL) to accomodate various flask sizes
Manufacturer:  Heidolph™ 015890280
 View more versions of this product
Catalog No. 05-404-055
Add to cart
Description
Description
MonoBlocks and PolyBlocks are component parts used in the assembly of Heidolph Starfish reaction stations
  • Used in conjunction with MonoBlock 5 x 250 mL and PolyBlock 1 x 250 mL only
  • Anodized aluminum for strong chemical resistance and fast heat-up times
Specifications
Specifications
25 mL
Monoblock and PolyBlock Used in The Assembly of Heidolph StarFish Reaction Station
Flask Insert