Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heidolph™ StarFish™ Work Station Accessories - Support Straps

Accessories for use with Heidolph StarFish™ Work Station
$93.00 - $410.00
Specifications
For Use With (Equipment) | Heidolph StarFish™ Work Station |
---|---|
Length (English) | 7.87 in. |
Length (Metric) | 200 mm |
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
05-404-150
![]() |
Heidolph™
015892180 |
Viton Support Strap |
Pack of 5 for $410.00
|
|
|||||
05-404-149
![]() |
Heidolph™
015892170 |
Silicone Support Strap |
Pack of 5 for $93.00
|
|
|||||
Description
Specifications
Heidolph StarFish™ Work Station | |
7.87 in. | |
200 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title