Learn More
Heidolph™ StarFish™ Work Station Accessories - Vials

Accessories for use with Heidolph StarFish™ Work Station
$308.00 - $1287.00
Catalog Number | Mfr. No. | Dimensions (L x W) | Vial Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Vial Volume (Metric) | Price | Quantity & Availability | ||||
05-404-128
![]() |
Heidolph™
015891010 |
21 x 70 mm | 14.7 mL |
Each for $862.00
|
|
||||
05-404-131
![]() |
Heidolph™
015891040 |
12 x 35 mm | 2 mL |
Each for $1,287.00
|
|
||||
05-404-130
![]() |
Heidolph™
015891030 |
15 x 45 mm | 3.7 mL |
Each for $308.00
|
|
||||
Description
Choice of size: 2mL 12 x 35 mm, 1 dram 15 x 45 mm, 2 dram 17 x 60 mm, and 4 dram 21 x 70 mm
Specifications
Heidolph StarFish™ Work Station | |
Vial |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.