Learn More
Heidolph™ Tornado Plus Overhead Stirring System, Stirrer Shafts
SDP
Stirrer shafts for use with Heidolph Tornado Plus
Supplier:  Heidolph™ 015861860
 View more versions of this product
Catalog No. 05-403-867
Add to Cart
Description
Description
A range of PTFE stirrer paddles have been designed specifically for the Tornado.
  • Centrifugal
  • Anchor
  • Propeller
The Centrifugal stirrer paddles will fit into any one of the standard narrow neck Carousel 6 Plus 50 mL, 100 mL, 170 mL or 250 mL flasks. The Anchor and Propeller stirrer paddles have been designed to fit in the wide neck 100 mL and 250 mL flasks.
Specifications
Specifications
Stirrer Shaft Assembly