Heidolph™ Tuttnauer™ Autoclaves Products for Science Education
Slow and fast exhaust in one unit.
$7918.00
Specifications
For Use With (Application) Sterilization of liquids, media, instruments, glassware, clothing and waste
Heating Time 20 min.
View More Specs
  • Heat-insulated, double-locking door protects against accidental burns; the door cannot be opened while the chamber is under pressure
  • Automatic low water level shut-off prevents damage to the heater elements if there is insufficient water for the sterilization cycle
  • Over-temperature cut-off and over-pressure valve ensure safe operation
  • UL- and CSA- approved
Economical Analog Sterilizers
  • These economical, manually operated models feature analog controls and gauges for simple operation
  • 60-minute cycle timer
  • Temperature selection from 1000°C to 1340°C
  • Combination pressure and temperature gauge with peak indicator
Warranty
Two years
Specifications
Sterilization of liquids, media, instruments, glassware, clothing and waste
20 min.
  • Products 2
  • Description
  • Specifications
  • Products 2
    Catalog Number Mfr. No. Capacity Diameter (English) Chamber Diameter (Metric) Chamber Model Price Quantity & Availability  
    S63039Products for Science Education Accessories Heidolph North America
    023210100/EMD
    9 in. 22.8cm 2340M, 120V Each for $7,918.00
    Check Availability
    Add to Cart
    S63100AProducts for Science Education Heidolph North America
    023210401/EMD
    6 gal., 23L 10 in. 25.4cm 2540E N/A N/A N/A  
    This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Description
    Specifications
    Related Products