Learn More
Heidolph™ Heidolph™ Tuttnauer™ Benchtop Sterilizers: M Series Available on GSA/VA Contract for Federal Government customers only.
Compact sterilizers featuring analog controls that provide a fast, safe, and convenient way to sterilize both solids and liquids
Manufacturer:  Heidolph™ 023210631
 View more versions of this product
Catalog No. 02-964-40
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
Economical analog controls and gauges for simple operation 60-minute cycle timer. Temperature selection from 100°C to 134°C (212°F to 273°F). Combination pressure and temperature gauge with peak indicator.
Standard Operating Features
  • Self-contained design eliminates external plumbing and hardwiring for easy installation
  • Simple operation
  • ASME-stamped, Type 316 L stainless-steel chamber ensures long-lasting reliability, durability, and high-corrosion resistance
  • Fast/slow exhause (fast allows quick cycle time of solids while slow exhause prevents media and liquid boil-over)
  • Closed-door drying system maintains sterility
  • Drying cycle removes residual moisture after sterilization
  • Continuous pressure purge eliminates air pockets and ensures thorough, even sterilization
  • Large-capacity reservoir provides ample water supply to avoid frequent fill-ups
  • Water drain is conveniently located on top to allow quick and easy cleaning of reservoir
  • Automatic shut-off feature saves energy
Standard Safety Features
  • Heat-insulated, double-locking door protects against accidental burns
  • Door cannot be opened while chamber is under pressure
  • Automatic low water level shut-off prevents damage to the heater elements if there is insufficient water for the sterilization cycyle
  • Overtemperature cutoff and overpressure valve ensure safe operation
Sterilization of liquids, media, instruments, glassware, clothing, and waste
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Service Plan
Service agreement available.
Certifications
UL listed, CSA, ASME (chamber)
Warranty and Services
One year
Specifications
Specifications
7.8 gal., 34.4L
UL, CSA, ASME, U.S. EPA
54.6cm
16.1 in.
34.4L
12.2 in.
31cm
18 in.
3545M
212 to 273°F
60kg
30 min.
2.5cm
50/60Hz
No
220V
7.8 in.
69.7cm
13.6A
21.5 in.
40.8cm
7.8 gal.
46.9cm
18.5 in.
Sterilization of liquids, media, instruments, glassware, clothing and waste
54.6cm
100 to 134°C
132 lb.
220V 50/60Hz
1 in.
Analog controls
No
2.0
2200w
23.5 in.
19.8cm
Documents
Accessories