Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heidolph™ Tuttnauer™ Vertical Top-Loading Autoclaves Accessories - Baskets
Ideal for medical waste sterilization
Supplier: Heidolph™ 023211840
Description
Choices include wire or solid baskets
Specifications
14.06 in. | |
Clinical and Medical Lab Instrument | |
13 in. | |
3850, 3870 | |
Large |
35.71 cm | |
Heidolph Tuttnauer Vertical Top Loading Autoclave | |
33.02 cm | |
Basket |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction