Heidolph™ Tuttnauer™ Vertical Top-Loading Autoclaves Accessories - Baskets Available on GSA/VA Contract for Federal Government customers only.

Ideal for medical waste sterilization

Manufacturer: Heidolph North America 023211800

 View more versions of this product

Catalog No. 13-889-486

  Check Availability
Add to Cart

    Specifications

    Diameter (English) Outside 8.94 in.
    Diameter (Metric) Outside 22.7cm
    For Use With (Application) Clinical and medical lab instrument
    For Use With (Equipment) Heidolph tuttnauer vertical top loading autoclave
    Height (English) 13.78 in.
    Height (Metric) 35cm
    Model 2540
    Size Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit