Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Helicobacter pylori Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Helicobacter pylori, Rabbit Polyclonal Antibody.

$216.00 - $1073.00

Specifications

Antigen Helicobacter pylori
Clone Polyclonal
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
RB9070P0
View Documents
Thermo Scientific™
RB9070P0A
100μL Each for $216.00
Add to cart
 
RB9070P1
View Documents
Thermo Scientific™
RB9070P1A
500μL Each for $485.00
Add to cart
 
RB9070P
View Documents
Thermo Scientific™
RB9070PA
1mL Each for $1,073.00
Add to cart
 
RB9070R7
View Documents
Thermo Scientific™
RB9070R7A
7mL Each for $305.50
Add to cart
 
RB9070RQ
View Documents
Thermo Scientific™
RB9070RQA
12mL Each for $508.50
Add to cart
 
Description

Description

Obtain accurate, reproducible results in Immunohistochemistry experiments with Thermo Scientific™ Helicobacter pylori, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Helicobacter pylori is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Helicobacter pylori

Epitope: Not determined

Immunogen: Purified Helicobacter pylori

Positive Control: Helicobacter pylori infected tissue

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Helicobacter pylori
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
Polyclonal
Polyclonal
Rabbit
Infectious Disease
Purified Helicobacter pylori
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit