Learn More
Helena Laboratories™ ColoCARE™ FOB Test Kit
Safer, easier and more pleasant to use than traditional guaiac slide tests. Simply place a ColoCARE test pad in the toilet after a bowel movement, watch for a color change and then flush the pad away.
Specifications
For Use With (Application) To test fecal occult blood
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Includes Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Includes Product Type Price Quantity & Availability  
23-038-045
View Documents
Helena Laboratories™
5656
100 Pack 100 ColoCARE Tests, instructions for medical personnel Hospital pack
Pack of 100 for $347.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-038-043
View Documents
Helena Laboratories™
5651
50 Pack 50 Patient kits, each containing 3 ColoCARE Tests, 1 patient instruction sheet, 1 ColoCARE reply card, 50 “ColoCARE Dispensed Today” labels, instructions Office pack
Pack of 50 for $376.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • CLIA Complexity: Waived
  • Throw-in-the-bowl test for detecting pre-symptomatic occult bleeding caused by gastrointestinal diseases
  • Clean and disposable, easy for elderly patients to see and interpret, and extremely sensitive, with no increase in the false positive rate
  • Requires no stool handling, no chemical developers, no laboratory processing, and no mailing of biohazards
  • Test pad consists of biodegradable paper chemically treated with a chromogen; floated on the water surface in the toilet bowl
  • With three reaction sites: a large test square and two smaller control squares to verify the system functions properly
Order Info
Office Pack for physician's office contains 50 patient kits, each including three ColoCARE tests, patient instruction sheet, ColoCARE reply card, 50 “ ColoCARE Dispensed Today” labels, and instructions for medical personnel; Hospital Pack for hospital laboratory or nurse's station contains 100 ColoCARE tests and instructions for medical personnel
Specifications
Specifications
To test fecal occult blood