Learn More
Helena Laboratories™ ColoCARE™ FOB Test Kit
Safer, easier and more pleasant to use than traditional guaiac slide tests. Simply place a ColoCARE test pad in the toilet after a bowel movement, watch for a color change and then flush the pad away.
Manufacturer:  Helena Laboratories™ 5656
 View more versions of this product
Catalog No. 23-038-045
  Check Availability
Add to cart
Specifications
Description
  • CLIA Complexity: Waived
  • Throw-in-the-bowl test for detecting pre-symptomatic occult bleeding caused by gastrointestinal diseases
  • Clean and disposable, easy for elderly patients to see and interpret, and extremely sensitive, with no increase in the false positive rate
  • Requires no stool handling, no chemical developers, no laboratory processing, and no mailing of biohazards
  • Test pad consists of biodegradable paper chemically treated with a chromogen; floated on the water surface in the toilet bowl
  • If detectable blood is present, the hemoglobin reacts with the chromogen, and a blue and/or green color reaction occurs
  • With three reaction sites: a large test square and two smaller control squares to verify the system functions properly
Description & Specifications
Specifications
Waived
100 ColoCARE Tests, instructions for medical personnel
Hospital pack
To test fecal occult blood
100 Pack