Helena Laboratories™ ColoScreen™ FOB Test Kit Available on GSA/VA Contract for Federal Government customers only.
Convenient triple slide provides three successive tests in one unit
Manufacturer: Helena Laboratories™ 5083
 View more versions of this product
Catalog No. 23-038-048
  Check Availability
Add to Cart
    Specifications
    Product Type ColoScreen Clinic Pack
    For Use With (Application) Fecal Occult Blood Testing
    Includes 10 x 34 Triple unit ColoScreen slides with on-slide, ColoCheck monitors, 20 x 15mL ColoScreen developer, 1000 specimen applicator
    Quantity 1000 Pack
    Clia Complexity Waived
    • CLIA Complexity: Waived
    • Ideal for physician's office or clinic use
    • Classic guaiac slide format
    • Office Pack allows slides, sample applicators and mailing envelopes to be conveniently distributed to patients, who can then prepare test slides in the privacy of their homes
    • Clinic Pack is designed for labs preferring the triple unit slides
    • Lab Pack is provided in order that each laboratory can stock the volume that best fits its demands
    • Each single slide in the triple unit has two test areas so that multiple sites from each specimen may be tested, increasing accuracy
  • Description & Specifications
Related Products
Product Content Feedback