Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Mouse anti-Bacteria, Clone: HP/1335, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30766420UG
Description
Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Helicobacter Pylori | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
H. pylori | |
Total sonicate of Helicobacter pylori | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunofluorescence | |
HP/1335 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Bacteria | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction