Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Mouse anti-Bacteria, Clone: HP/1335, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | Helicobacter Pylori |
---|---|
Clone | HP/1335 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
Applications | Flow Cytometry, ELISA, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122802
|
Novus Biologicals
NBP30766420UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
Helicobacter Pylori Monoclonal specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Helicobacter Pylori | |
0.2 mg/ml | |
Flow Cytometry, ELISA, Immunofluorescence | |
Unconjugated | |
Mouse | |
Bacteria | |
H. pylori | |
IgG1 κ | |
Protein A or G purified |
HP/1335 | |
Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
Total sonicate of Helicobacter pylori | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title