Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter Pylori Mouse anti-Bacteria, Clone: HP/1336, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30767410UG

 View more versions of this product

Catalog No. NB122821


Add to cart

Description

Description

Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in Bacteria samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications

Specifications

Helicobacter Pylori
HP/1336
Unconjugated
10 mM PBS with 0.05% BSA
H. pylori
Total sonicate of Helicobacter pylori
Protein A or G purified
RUO
Primary
Bacteria
Flow Cytometry, ELISA, Immunofluorescence
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Store at 4°C.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit