Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Helicobacter Pylori Mouse anti-H. Pylori, DyLight 650, Clone: HP/1334, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314092C
This item is not returnable.
View return policy
Description
Helicobacter Pylori Monoclonal antibody specifically detects Helicobacter Pylori in H. Pylori samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Helicobacter Pylori | |
Monoclonal | |
DyLight 650 | |
H. Pylori | |
Total sonicate of Helicobacter pylori | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry, ELISA, Immunofluorescence | |
HP/1334 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
H. Pylori | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title