Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helicobacter pylori urease B Mouse anti-Bacteria, DyLight 550, Clone: 962, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242852R

Catalog No. NBP242852R


Add to cart

Description

Description

Helicobacter pylori urease B Monoclonal antibody specifically detects Helicobacter pylori urease B in Bacteria samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Helicobacter pylori urease B
962
Western Blot, ELISA
Purified
The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Western Blot, ELISA
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2a
0.1 ml
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit