Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Helios Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17970320UL

 View more versions of this product

Catalog No. NBP17970320


Add to cart

Description

Description

Helios Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Helios
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
IKZF2
Synthetic peptide directed towards the middle region of human IKZF2. Peptide sequence EREASPSNSCLDSTDSESSHDDHQSYQGHPALNPKRKQSPAYMKEDVKAL.
59 kDa
20ul
Adaptive Immunity, Immunology
Primary
22807
Western Blot
Western Blot 1:1000
NP_057344
HELIOS, IKAROS family zinc finger 2 (Helios), Ikaros family zinc finger protein 2, MGC34330, subfamily 1A, 2 (Helios), zinc finger DNA binding protein Helios, zinc finger protein Helios, ZNF1A2, ZNFN1A2
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit