Learn More
Microbiologics™ Helix Elite™ BD MAX™ Enteric Parasite Verification Panel
SDP
Features the manufacturer recommended QC strains in an easy-to-use, room-temperature stable format
Supplier:  Microbiologics™ 8182
Catalog No. 23-004-140
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative. View substitute products »
Includes: Each kit includes five vials of dried synthetic DNA and one vial of molecular standard water for rehydration. Each vial is comprised of pooled Cryptosporidium spp., Entamoeba histolytica, and Giardia lamblia synthetic DNA.
This item is not returnable. View return policy
Description
Description
Biosafety Level: 1
Specifications
Specifications
1
Helix Elite BD MAX Enteric Parasite Verification Panel
Cryptosporidium spp., Entamoeba histolytica, Giardia lamblia
Drinking Water Testing
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Validated with BD MAX Enteric Parasite Panel; Formerly BD MAX Enteric Parasite Control Panel. For in vitro diagnostic use. International restriction might apply.