Learn More
MCR Safety HellKat 2 MAX6 Lens
Click to view available options
Lens Tint:
Amber
Blue
Brown
Clear
Gray
Description
- Lightweight, contoured frameless design
- MAX6™ premium anti-fog lens coating
- Excellent orbital seal for eye protection
- Non-slip soft bayonet temple
- Soft, secure TPR nose pad
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Lens Tint | Amber |
For Use With (Application) | Manufacturing |
Frame Color | Amber |
No. per Pack | 1 Pair |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |